Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methoxyflavone | 7245-02-5 | 99.9% | 252.26 | 1 ML
SDP

Catalog No. 5000425369
Encompass_Preferred

3-Methoxyflavone is a flavonoid compound isolated from Artemisia incanescens with antiviral activity. It is supplied as a 10 mM solution in 1 mL of DMSO, suitable for research applications.

  • Flavonoid compound
  • Exhibits antiviral activity
  • Purity of 99.9%
  • Provided as a 10 mM solution in DMSO
  • Recommended storage for solution: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-25369 Supplier Medchemexpress LLC Supplier No. HYN949910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methoxyflavone is a flavonoid compound isolated from Artemisia incanescens with antiviral activity. It is supplied as a 10 mM solution in 1 mL of DMSO, suitable for research applications.

  • Flavonoid compound
  • Exhibits antiviral activity
  • Purity of 99.9%
  • Provided as a 10 mM solution in DMSO
  • Recommended storage for solution: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.