Learn More
Medchemexpress LLC 3-Methoxyflavone | 7245-02-5 | 99.9% | 252.26 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN949950MG
Encompass_Preferred
3-Methoxyflavone is a flavonoid compound that has been isolated from Artemisia incanescens. It exhibits antiviral activity and can inhibit human BCRP (Breast Cancer Resistance Protein) expressed in MDCK2 cells. It is available as an off-white to light yellow solid.
  • Isolated from Artemisia incanescens.
  • Exhibits antiviral activity.
  • Inhibits human BCRP expressed in MDCK2 cells.
  • Available as an off-white to light yellow solid.
Catalog No. 50-004-26084