Learn More
Medchemexpress LLC 3-Methoxyflavone | 7245-02-5 | MFCD00017612 | 99.88% | 252.26 | 100 MG
SDP
Catalog No. 5000425584
Encompass_Preferred
3-Methoxyflavone is a flavonoid compound isolated from Artemisia incanescens with antiviral activity. It is supplied as an off-white to light yellow solid.
  • Isolated from Artemisia incanescens
  • Exhibits antiviral activity
  • High purity of 99.88%
  • Available as a solid, off-white to light yellow in color
Catalog No. 50-004-25584 Supplier Medchemexpress LLC Supplier No. HYN9499100MG
3-Methoxyflavone is a flavonoid compound isolated from Artemisia incanescens with antiviral activity. It is supplied as an off-white to light yellow solid.
  • Isolated from Artemisia incanescens
  • Exhibits antiviral activity
  • High purity of 99.88%
  • Available as a solid, off-white to light yellow in color