Learn More
Medchemexpress LLC HY-N6629 50mg Medchemexpress, 3'-Methoxyflavonol CAS:76666-32-5 Purity:>98%
SDP
Catalog No. 501933963
Encompass_Preferred
Medchemexpress, HY-N6629 50mg 3'-Methoxyflavonol CAS:76666-32-5 3'-Methoxyflavonol is a selective agonist of neuromedin U 2 receptor ( NMU2R ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3963 Supplier Medchemexpress LLC Supplier No. HYN662950MG
Medchemexpress, HY-N6629 50mg 3'-Methoxyflavonol CAS:76666-32-5 3'-Methoxyflavonol is a selective agonist of neuromedin U 2 receptor ( NMU2R ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.