Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methoxyisoquinoline-1-carboxylic acid | 374917-64-3 | MFCD01845410 | 99.6% | 203.20 g/mol | C11H9NO3 | 250 MG
SDP

Catalog No. 5000209415
Encompass_Preferred

3-Methoxyisoquinoline-1-carboxylic acid is an organic drug intermediate used in medicinal chemistry for synthesis of substituted isoquinoline derivatives. It is supplied as a solid and characterized by analytical data supporting high purity and structural identity.

  • used as a drug intermediate in chemical synthesis
  • white to yellow solid appearance
  • high purity: 99.6% (HPLC, per COA)
  • molecular formula: C11H9NO3
  • molecular weight: 203.20 g/mol
  • recommended storage: store at room temperature (stable 3 years); in solvent: -80°C (2 years), -20°C (1 year)
  • typical available quantity: 250 mg

Catalog No. 50-002-09415 Supplier Medchemexpress LLC Supplier No. 250MGHYW065670
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methoxyisoquinoline-1-carboxylic acid is an organic drug intermediate used in medicinal chemistry for synthesis of substituted isoquinoline derivatives. It is supplied as a solid and characterized by analytical data supporting high purity and structural identity.

  • used as a drug intermediate in chemical synthesis
  • white to yellow solid appearance
  • high purity: 99.6% (HPLC, per COA)
  • molecular formula: C11H9NO3
  • molecular weight: 203.20 g/mol
  • recommended storage: store at room temperature (stable 3 years); in solvent: -80°C (2 years), -20°C (1 year)
  • typical available quantity: 250 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.