Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenol, ≥96.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019737 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9737 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9737 Supplier MilliporeSigma Supelco Supplier No. 447771ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

CAS 150-19-6
Density 1.131 g/mL (at 25°C)
Linear Formula CH3 OC6H4 OH
MDL Number MFCD00002267
Boiling Point 113°C to 115°C (5 mmHg)
Synonym 3-Hydroxyanisol; Resorcinol monomethyl ether
Type Analytical Standard
Quantity 1 mL
Formula Weight 124.14
Percent Purity ≥96.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.