Learn More
Medchemexpress LLC 3-methoxyphenylacetic acid | 1798-09-0 | MFCD00004334 | 99.4% | 166.17 g/mol | C9H10O3 | 25 G
SDP
Catalog No. 5000303575
Encompass_Preferred
3-Methoxyphenylacetic acid is a small organic acid used as a research reagent, synthetic building block, and analytical standard in chemical and biochemical applications. Supplied as a solid powder, it is suitable for storage at low temperatures and is commonly used in organic synthesis, metabolite studies, and analytical method development.
  • Used as a building block in organic synthesis.
  • Suitable as an analytical standard for chromatography and spectrometry.
  • Supplied as a solid powder with multi-gram pack sizes.
  • High purity for reliable experimental results.
  • Stable under recommended cold storage conditions.
Catalog No. 50-003-03575 Supplier Medchemexpress LLC Supplier No. 25GHYW015343
Add to Cart
Add to Cart
3-Methoxyphenylacetic acid is a small organic acid used as a research reagent, synthetic building block, and analytical standard in chemical and biochemical applications. Supplied as a solid powder, it is suitable for storage at low temperatures and is commonly used in organic synthesis, metabolite studies, and analytical method development.
  • Used as a building block in organic synthesis.
  • Suitable as an analytical standard for chromatography and spectrometry.
  • Supplied as a solid powder with multi-gram pack sizes.
  • High purity for reliable experimental results.
  • Stable under recommended cold storage conditions.