Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methoxyphenylacetic acid | 1798-09-0 | MFCD00004334 | 99.6% | 166.17 g/mol | C9H10O3 | 50 G
SDP

Catalog No. 5000303263
Encompass_Preferred

3-methoxyphenylacetic acid is an organic reagent and reference standard supplied as a solid for laboratory research use. It is commonly used in organic synthesis and biochemical assays where a high-purity m-anisylacetic acid derivative is required.

  • High purity: 99.6% suitable for analytical and synthetic applications.
  • Molecular formula: C9H10O3.
  • Molecular weight: 166.17 g/mol.
  • Appearance: solid/powder for easy handling and weighing.
  • Storage stability: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in multiple solid pack sizes to support varying scale of experiments.

Catalog No. 50-003-03263 Supplier Medchemexpress LLC Supplier No. HYW01534350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-methoxyphenylacetic acid is an organic reagent and reference standard supplied as a solid for laboratory research use. It is commonly used in organic synthesis and biochemical assays where a high-purity m-anisylacetic acid derivative is required.

  • High purity: 99.6% suitable for analytical and synthetic applications.
  • Molecular formula: C9H10O3.
  • Molecular weight: 166.17 g/mol.
  • Appearance: solid/powder for easy handling and weighing.
  • Storage stability: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in multiple solid pack sizes to support varying scale of experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.