Learn More
Medchemexpress LLC 3-Methoxyphenylethylamine | 2039-67-0 | 99.4% | 151.21 | 50 G
SDP
Catalog No. 5000404420
Encompass_Preferred
3-Methoxyphenylethylamine is an organic compound utilized as an intermediate for pharmaceuticals, dyestuffs, and liquid crystal materials. It acts as a substrate for cytochrome P450 2D6 (CYP2D6), allowing its use in drug metabolism studies.
  • Serves as a model substrate in drug metabolism studies to investigate catalytic mechanisms and substrate selectivity.
  • Its oxidation product acts as a norepinephrine precursor.
  • Contributes to the nervous system.
Catalog No. 50-004-04420 Supplier Medchemexpress LLC Supplier No. HY4126850G
3-Methoxyphenylethylamine is an organic compound utilized as an intermediate for pharmaceuticals, dyestuffs, and liquid crystal materials. It acts as a substrate for cytochrome P450 2D6 (CYP2D6), allowing its use in drug metabolism studies.
  • Serves as a model substrate in drug metabolism studies to investigate catalytic mechanisms and substrate selectivity.
  • Its oxidation product acts as a norepinephrine precursor.
  • Contributes to the nervous system.