Learn More
Medchemexpress LLC 3β-methoxypregn-5-en-20-one | 511-26-2 | 99.7% | 330.50 g·mol⁻¹ | C22H34O2 | 10 MG
SDP
Catalog No. 5000239818
Encompass_Preferred
MAP4343 (3β-methoxypregnenolone, CAS 511-26-2) is a pregnenolone derivative used as a research reagent that modulates microtubule dynamics. It is supplied as a high-purity solid for in vitro and in vivo neurobiology studies, with documented solubility and handling guidance.
  • High purity solid suitable for research use.
  • Binds microtubule-associated protein 2 (MAP2) and stimulates tubulin polymerization.
  • Shows neuroprotective and behavioral effects in preclinical models.
  • Soluble in DMSO; in vivo formulation protocols described for dosing.
  • Provided with supporting documentation including COA and SDS.
Catalog No. 50-002-39818 Supplier Medchemexpress LLC Supplier No. HY10711610MG
MAP4343 (3β-methoxypregnenolone, CAS 511-26-2) is a pregnenolone derivative used as a research reagent that modulates microtubule dynamics. It is supplied as a high-purity solid for in vitro and in vivo neurobiology studies, with documented solubility and handling guidance.
  • High purity solid suitable for research use.
  • Binds microtubule-associated protein 2 (MAP2) and stimulates tubulin polymerization.
  • Shows neuroprotective and behavioral effects in preclinical models.
  • Soluble in DMSO; in vivo formulation protocols described for dosing.
  • Provided with supporting documentation including COA and SDS.