Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3β-methoxypregn-5-en-20-one | 511-26-2 | 99.7% | 330.50 g·mol⁻¹ | C22H34O2 | 100 MG
SDP

Catalog No. 5000239819
Encompass_Preferred

MAP4343 (3β-methoxypregnenolone) is a synthetic pregnenolone derivative that binds microtubule-associated protein 2 (MAP2), promotes tubulin polymerization, enhances neurite extension, and shows neuroprotective and antidepressant effects in preclinical studies. It is supplied as a high-purity research reagent for neurobiology and microtubule-related assays.

  • Chemical identity: 3β-methoxypregn-5-en-20-one (CAS 511-26-2).
  • Purity: 99.7%.
  • Molecular formula C22H34O2 and molecular weight 330.50 g·mol⁻¹.
  • Typical applications include tubulin polymerization assays and neurite outgrowth studies.
  • Provides neuroprotective and mood-related activity in preclinical models.
  • Available in solid and solution formats with laboratory-scale pack sizes.

Catalog No. 50-002-39819 Supplier Medchemexpress LLC Supplier No. HY107116100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAP4343 (3β-methoxypregnenolone) is a synthetic pregnenolone derivative that binds microtubule-associated protein 2 (MAP2), promotes tubulin polymerization, enhances neurite extension, and shows neuroprotective and antidepressant effects in preclinical studies. It is supplied as a high-purity research reagent for neurobiology and microtubule-related assays.

  • Chemical identity: 3β-methoxypregn-5-en-20-one (CAS 511-26-2).
  • Purity: 99.7%.
  • Molecular formula C22H34O2 and molecular weight 330.50 g·mol⁻¹.
  • Typical applications include tubulin polymerization assays and neurite outgrowth studies.
  • Provides neuroprotective and mood-related activity in preclinical models.
  • Available in solid and solution formats with laboratory-scale pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.