Learn More
Medchemexpress LLC 3β-methoxypregnenolone | 511-26-2 | 99.7% | 330.50 g·mol-1 | C22H34O2 | 50 MG
SDP
Catalog No. 5000239938
Encompass_Preferred
MAP4343 (3β-methoxypregnenolone) is a synthetic pregnenolone derivative used as a research reagent. It binds microtubule-associated protein 2 (MAP2), promotes tubulin polymerization, enhances neurite outgrowth, and shows neuroprotective activity in vitro. It is supplied as a solid and is commonly used in neurobiology and cell biology studies.
  • Modulates microtubule-associated protein 2 (MAP2) activity.
  • Promotes tubulin polymerization in vitro.
  • Enhances neurite outgrowth in neuronal cultures.
  • Provides neuroprotective effects against neurotoxic agents.
  • Available as a solid and as prepared solutions for cell studies.
  • Reported high purity suitable for research applications.
Catalog No. 50-002-39938 Supplier Medchemexpress LLC Supplier No. HY10711650MG
MAP4343 (3β-methoxypregnenolone) is a synthetic pregnenolone derivative used as a research reagent. It binds microtubule-associated protein 2 (MAP2), promotes tubulin polymerization, enhances neurite outgrowth, and shows neuroprotective activity in vitro. It is supplied as a solid and is commonly used in neurobiology and cell biology studies.
  • Modulates microtubule-associated protein 2 (MAP2) activity.
  • Promotes tubulin polymerization in vitro.
  • Enhances neurite outgrowth in neuronal cultures.
  • Provides neuroprotective effects against neurotoxic agents.
  • Available as a solid and as prepared solutions for cell studies.
  • Reported high purity suitable for research applications.