Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 3-Methoxypropylamine, 99%

Catalog No. AAB24095AU
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB24095AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB24095AU Supplier Thermo Scientific™ Supplier No. B24095AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5332-73-0 | C4H11NO | 89.14 g/mol

TRUSTED_SUSTAINABILITY

Specifications

CAS 5332-73-0
Melting Point <-70°
Density 0.872
Boiling Point 117-119°
Flash Point 27°(81°F)
Molecular Formula C4H11NO
MDL Number MFCD00014831
Quantity 1000 mL
UN Number UN2734
Synonym 1-Amino-3-methoxypropane;
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
SMILES COCCCN
IUPAC Name 3-methoxypropan-1-amine
Molecular Weight (g/mol) 89.14
Formula Weight 89.14
Percent Purity 99%
Physical Form Liquid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 226-241-3
RTECSNumber UI3335000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.