Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxythiophenol, 97%

Catalog No. p-7080912
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1064803 1 g
AAA1064806 5 g
AAA1064814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1064803 Supplier Thermo Scientific Chemicals Supplier No. A1064803
Only null left
Add to Cart

CAS: 15570-12-4 | C7H8OS | 140.2 g/mol

3-Methoxythiophenol is used in the preparation of allenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophenol is used in the preparation of allenes.

Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.

Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15570-12-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004841
InChI Key QMVAZEHZOPDGHA-UHFFFAOYSA-N
Synonym 3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol
PubChem CID 84989
IUPAC Name 3-methoxybenzenethiol
SMILES COC1=CC(=CC=C1)S

Specifications

Density 1.136
Boiling Point 225°C to 227°C
Flash Point 96°C (204°F)
Odor Unpleasant
Refractive Index 1.5865
Quantity 1 g
Beilstein 2041496
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in methanol,benzene,hexane,toluene and dichloromethane.
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 3-Methoxythiophenol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 239-617-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.