Learn More
Medchemexpress LLC 3-Methoxytyramine-d4 hydrochloride | 1216788-76-9 | MFCD06658564 | 98.0% | 207.69 g/mol | C9H10D4ClNO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY103638S1MG
Encompass_Preferred
3-Methoxytyramine-d4 hydrochloride is the deuterium-labeled (d4) hydrochloride salt of 3-methoxytyramine (3-O-methyl dopamine). It is provided as a solid reference standard for use as an internal standard or tracer in quantitative analytical methods.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Hydrochloride salt form for improved stability and handling.
  • Suitable for LC-MS, GC-MS, and NMR applications.
  • Available in small reference sizes (1 mg and 5 mg) for analytical workflows.
  • Useful for accurate quantification and method validation in bioanalysis.
Catalog No. 50-002-44897