Learn More
TARGETMOL CHEMICALS INC 3-Methoxytyramine hydrochlorid
SDP
Catalog No. 502878901
Encompass_Preferred
Also available in 1 mL, 100 mg, 250 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-Methoxytyramine hydrochloride (3-O-methyl Dopamine hydrochloride) is an inactive metabolite of dopamine which can activate trace amine associated receptor 1 (TAAR1). 3-methoxytyramine can be found primarily in human brain and most tissues tissues; and in blood, cerebrospinal fluid (csf) or urine. Within a cell, 3-methoxytyramine is primarily located in the cytoplasm. Purity 98.99%
Catalog No. 50-287-8901 Supplier TARGETMOL CHEMICALS INC Supplier No. T47101G
Also available in 1 mL, 100 mg, 250 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-Methoxytyramine hydrochloride (3-O-methyl Dopamine hydrochloride) is an inactive metabolite of dopamine which can activate trace amine associated receptor 1 (TAAR1). 3-methoxytyramine can be found primarily in human brain and most tissues tissues; and in blood, cerebrospinal fluid (csf) or urine. Within a cell, 3-methoxytyramine is primarily located in the cytoplasm. Purity 98.99%