Learn More
Medchemexpress LLC 3-methoxytyramine hydrochloride | 1477-68-5 | MFCD00012902 | 98.2% | 203.67 g·mol⁻¹ | C9H14ClNO2 | 1 ML
SDP
Catalog No. 5000246722
Encompass_Preferred
3-methoxytyramine hydrochloride is the hydrochloride salt of 3-methoxytyramine, an extracellular metabolite of dopamine that can activate trace amine-associated receptor 1 (TAAR1). It is supplied for research use in neurochemical and pharmacology studies, and as an analytical standard.
  • Hydrochloride salt of 3-methoxytyramine, a dopamine metabolite.
  • Supplied as a 10 mM solution, 1 mL vial.
  • Purity 98.2%.
  • Suitable for receptor pharmacology and neurochemistry research.
  • Available in solid and solution formats for flexibility in experimental workflows.
Catalog No. 50-002-46722 Supplier Medchemexpress LLC Supplier No. HY10363810MM 1ML
3-methoxytyramine hydrochloride is the hydrochloride salt of 3-methoxytyramine, an extracellular metabolite of dopamine that can activate trace amine-associated receptor 1 (TAAR1). It is supplied for research use in neurochemical and pharmacology studies, and as an analytical standard.
  • Hydrochloride salt of 3-methoxytyramine, a dopamine metabolite.
  • Supplied as a 10 mM solution, 1 mL vial.
  • Purity 98.2%.
  • Suitable for receptor pharmacology and neurochemistry research.
  • Available in solid and solution formats for flexibility in experimental workflows.