Learn More
Medchemexpress LLC 3-methoxytyramine hydrochloride | 1477-68-5 | MFCD00060621 | 98.2% | 203.67 g/mol | C9H14ClNO2 | 10 G
SDP
Catalog No. 5000245036
Encompass_Preferred
3-Methoxytyramine hydrochloride is the hydrochloride salt of 3-methoxytyramine, a dopamine metabolite that can activate trace amine-associated receptor 1 (TAAR1). Supplied as a solid research reagent and analytical standard, it is commonly used in neurochemical, metabolic, and pharmacology studies.
  • Used as an analytical standard for metabolite quantification.
  • Activates trace amine-associated receptor 1 in biological assays.
  • Applicable to neurological and metabolic research.
  • Provided as a hydrochloride salt with specified purity (≈98.2%).
  • Solid form with molecular weight 203.67 g/mol for the hydrochloride salt.
Catalog No. 50-002-45036 Supplier Medchemexpress LLC Supplier No. HY10363810G
3-Methoxytyramine hydrochloride is the hydrochloride salt of 3-methoxytyramine, a dopamine metabolite that can activate trace amine-associated receptor 1 (TAAR1). Supplied as a solid research reagent and analytical standard, it is commonly used in neurochemical, metabolic, and pharmacology studies.
  • Used as an analytical standard for metabolite quantification.
  • Activates trace amine-associated receptor 1 in biological assays.
  • Applicable to neurological and metabolic research.
  • Provided as a hydrochloride salt with specified purity (≈98.2%).
  • Solid form with molecular weight 203.67 g/mol for the hydrochloride salt.