Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxytyramine hydrochloride, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013160 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3160 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3160 Supplier MilliporeSigma Supelco Supplier No. 7502425MG
Add to Cart
Add to Cart

3-Methoxytyramine hydrochloride is a minor dopamine metabolite, found naturally in the cactus, Trichocereus pachanoi.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1477-68-5
Linear Formula CH3 OC6H3-4-(OH)CH2 CH2 NH2·HCl
MDL Number MFCD00012902
Melting Point 213°C to 215°C
Synonym 3-MT; 3-Methoxy-4-hydroxyphenethylamine hydrochloride; 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
Type Analytical Standard
Quantity 25 mg
Formula Weight 203.67
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.