Learn More
Cayman Chemical 3-Methoxytyrosine, Purity: ≥95%, CAS: 7636-26-2, 50 mg
SDP
Catalog No. 502913075
Encompass_Preferred
An active metabolite of L-DOPA; inhibits L-DOPA-induced neuroprotection in rat primary mesencephalic dopaminergic neurons at 10 and 100 µM
Catalog No. 50-291-3075 Supplier Cayman Chemical Supplier No. 3644350 MG
An active metabolite of L-DOPA; inhibits L-DOPA-induced neuroprotection in rat primary mesencephalic dopaminergic neurons at 10 and 100 µM