Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1,5-pentanediol, 99%

Catalog No. AAB2054606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2054606 5 g
AAB2054614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2054606 Supplier Thermo Scientific Chemicals Supplier No. B2054606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4457-71-0 | C6H14O2 | 118.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4457-71-0
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00039547
InChI Key SXFJDZNJHVPHPH-UHFFFAOYSA-N
Synonym 3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference
PubChem CID 20524
IUPAC Name 3-methylpentane-1,5-diol
SMILES CC(CCO)CCO

Specifications

Density 0.97
Flash Point 143°C (289°F)
Refractive Index 1.454
Quantity 5 g
Beilstein 1697331
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material 3-Methyl-1,5-pentanediol
EINECSNumber 224-709-1
RTECSNumber SA0800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.