Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methyl-1-oxo-1,5-dihydropyrido[1,2-a]benzimidazole-4-carbonitrile

Catalog No. OTV0005951G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0005951G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0005951G Supplier Sigma Aldrich Supplier No. OTV0005951G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Irradiation of cyclohexene-1-carbonitrile with the full arc of hanovia 450-w lamp yields bicycle[3.1.0]hexane-1-carbonitrile
  • Photochemistry of cyclohexene-1-carbonitrile has been investigated.cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid

Specifications

CAS 60792-57-6
Molecular Formula C13H9N3O
Linear Formula C13H9N3O
MDL Number MFCD00649384
Quantity 10 mL
Molecular Weight (g/mol) 223.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.