Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-p-tolyl-5-pyrazolone 98.0+%, TCI America™
SDP

Catalog No. M055425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M055425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M055425G Supplier TCI America Supplier No. M055425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86-92-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00035708
InChI Key IOQOLGUXWSBWHR-UHFFFAOYSA-N
PubChem CID 66591
IUPAC Name 3-methyl-1-(4-methylphenyl)-4,5-dihydro-1H-pyrazol-5-one
SMILES CC1=NN(C(=O)C1)C1=CC=C(C)C=C1

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 25 g
Formula Weight 188.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1-p-tolyl-5-pyrazolone
RTECSNumber UQ9680000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.