Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Castor oil, specified according to the requirements of Ph.Eur.

Catalog No. AC348850025
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle

Catalog No. AC348850025

Supplier: Thermo Scientific Chemicals 348850025

Only null left
Add to Cart

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
MDL Number MFCD00130746
InChI Key ZEMPKEQAKRGZGQ-AAKVHIHISA-N
Synonym castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol
PubChem CID 14030006
IUPAC Name 2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O

Specifications

Acidity 2.0mg KOH/g max.
Absorbance 1.5 max. (at the absorption maximum at 269 +/- 1nm)
Melting Point -10.0°C
Color Colorless or Yellow
Density 0.9570g/mL
Boiling Point 229.0°C
Flash Point 229°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4770 to 1.4810
Quantity 2.5 L
Merck Index 14,1898
Specific Gravity 0.957
Specific Rotation Condition + 3.50 (20.00°C c=neat)
Specific Rotation + 3.50
Solubility Information Solubility in water: .. Other solubilities: miscible with alcohol, benzene, chloroform,, carbondisulfide, ether and glacial acetic acid
Viscosity 0,6-0,8 Pas (25°C)
Grade Eu. Ph.
Concentration or Composition (by Analyte or Components) of fatty acids (GC): Passes Test (per Eur. Pharm.)
Physical Form Viscous Hygroscopic Liquid
Chemical Name or Material Castor oil
Show More Show Less
EINECSNumber 232-293-8
RTECSNumber FI4100000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.