Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methyl-2(5H)-furanone

Catalog No. 3935095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-350-95G 5 g
39-350-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-350-95G Supplier Sigma Aldrich Supplier No. 3935095G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methyl-2(5h)-furanone, a five membered conjugated lactone, is a volatile compound
  • Its synthesis has been reported
  • It is reported to be one of the predominant constituent of the flavor of pandan leaves
  • It is also found in thai soy sauce, arabica roasted coffee samples from brazil, bio-oil from pine wood sawdust.3-methyl-2(5h)-furanone is the suitable model compound used to investigate the fragmentation mechanism of five membered lactones by electrospray ionisation tandem mass spectrometry and also to study the airway irritation of isoprene, isoprene/ozone, and isoprene/ozone/nitrogen dioxide mixture

Specifications

CAS 22122-36-7
Density 1.13 g/mL (at 25°C (literature))
Boiling Point 97°C to 99°C (20 mmHg)
Molecular Formula C5H6O2
Refractive Index n20/D 1.467 (literature)
Linear Formula C5H6O2
MDL Number MFCD00191545
Quantity 5 g
Molecular Weight (g/mol) 98.1
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.