Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 99.9%, MP Biomedicals™

Catalog No. ICN15163801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15163801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15163801 Supplier MP Biomedicals, Inc Supplier No. 0215163801

missing translation for 'validMainframeMsg'

Soluble in methanol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14448-67-0
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.699
MDL Number MFCD00149370
InChI Key OEZPVSPULCMUQB-DQMXGCRQSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
PubChem CID 9575839
IUPAC Name (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.Cl

Specifications

Color Tan
Quantity 1 g
Formula Weight 233.7
Physical Form Powder
Chemical Name or Material 3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.