Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methyl-2-Buten-2-Ylboronic Acid Pinacol Ester

Catalog No. 6882901G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-829-01G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 68-829-01G Supplier Sigma Aldrich Supplier No. 6882901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Methyl-2-Buten-2-Ylboronic Acid Pinacol Ester

Specifications

CAS 163105-90-6
Melting Point 114°C to 119°C
Molecular Formula C30H47O2P
Refractive Index n20/D 1.450
Linear Formula C30H47O2P
MDL Number MFCD08458882
Quantity 100 mg
Molecular Weight (g/mol) 470.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.