Learn More
Medchemexpress LLC 3-Methyl-2-butenal | 107-86-8 | 97.3% | 84.12 | 10 G
SDP
Catalog No. 5000404154
Encompass_Preferred
3-Methyl-2-butenal, also known as 3-Methylbut-2-en-1-al, can be isolated from hops. This product is intended for research use only.
  • Appearance: Liquid (density: 0.9 g/cm³)
  • Color: Colorless to light yellow
  • Initial source: Plants Moraceae Humulus lupulus L. (hops)
Catalog No. 50-004-04154 Supplier Medchemexpress LLC Supplier No. HY3440910G
3-Methyl-2-butenal, also known as 3-Methylbut-2-en-1-al, can be isolated from hops. This product is intended for research use only.
  • Appearance: Liquid (density: 0.9 g/cm³)
  • Color: Colorless to light yellow
  • Initial source: Plants Moraceae Humulus lupulus L. (hops)