Learn More
Medchemexpress LLC 3-Methyl-2-butenal | 107-86-8 | 97.3% | 84.12 | 500 G
SDP
Catalog No. 5000404459
Encompass_Preferred
3-Methyl-2-butenal (3-Methylbut-2-en-1-al) is a compound that can be isolated from hops. It is intended for research use only and is not sold to patients. It appears as a liquid with a density of 0.9 g/cm³, and its color can range from colorless to light yellow. Its initial source is plants, specifically Moraceae Humulus lupulus L.
  • Can be isolated from hops
  • For research use only
  • Appears as a liquid
  • Density of 0.9 g/cm³
  • Colorless to light yellow liquid
  • Initial source from plants (Moraceae Humulus lupulus L.)
Catalog No. 50-004-04459 Supplier Medchemexpress LLC Supplier No. HY34409500G
3-Methyl-2-butenal (3-Methylbut-2-en-1-al) is a compound that can be isolated from hops. It is intended for research use only and is not sold to patients. It appears as a liquid with a density of 0.9 g/cm³, and its color can range from colorless to light yellow. Its initial source is plants, specifically Moraceae Humulus lupulus L.
  • Can be isolated from hops
  • For research use only
  • Appears as a liquid
  • Density of 0.9 g/cm³
  • Colorless to light yellow liquid
  • Initial source from plants (Moraceae Humulus lupulus L.)