Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-butenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A114825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A114825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A114825ML Supplier TCI America Supplier No. A114825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1191-16-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00036569
InChI Key XXIKYCPRDXIMQM-UHFFFAOYSA-N
Synonym prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate
PubChem CID 14489
IUPAC Name 3-methylbut-2-en-1-yl acetate
SMILES CC(C)=CCOC(C)=O

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butenyl Acetate
RTECSNumber EM9473700
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.