Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-butenyl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B140925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B140925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B140925G Supplier TCI America Supplier No. B140925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5205-11-8
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00036570
InChI Key INVWRXWYYVMFQC-UHFFFAOYSA-N
Synonym Benzoic Acid 3-Methyl-2-butenyl Ester, Benzoic Acid Prenyl Ester, Prenyl Benzoate
PubChem CID 21265
IUPAC Name 3-methylbut-2-enyl benzoate
SMILES CC(=CCOC(=O)C1=CC=CC=C1)C

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 g
Formula Weight 190.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butenyl Benzoate
EINECSNumber (3)-1357
RTECSNumber EM9473900
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.