Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methyl-2-cyclopenten-1-one | 2758-18-1 | 99.1% | 96.13 | 1 ML
SDP

Catalog No. 5000452171
Encompass_Preferred

3-Methyl-2-cyclopenten-1-one is an endogenous metabolite for research use only, not intended for patient use.

  • Endogenous metabolite
  • Purity of 99.12%
  • Molecular weight of 96.13
  • Chemical formula C6H8O
  • Liquid appearance, light yellow to brown
  • Soluble in DMSO

Catalog No. 50-004-52171 Supplier Medchemexpress LLC Supplier No. HYW01301410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methyl-2-cyclopenten-1-one is an endogenous metabolite for research use only, not intended for patient use.

  • Endogenous metabolite
  • Purity of 99.12%
  • Molecular weight of 96.13
  • Chemical formula C6H8O
  • Liquid appearance, light yellow to brown
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.