Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methyl-2-cyclopenten-1-one | 2758-18-1 | 99.1% | 96.13 | 5 G
SDP

Catalog No. 5000454093
Encompass_Preferred

3-Methyl-2-cyclopenten-1-one is an endogenous metabolite.

  • Endogenous metabolite
  • Appearance: liquid (density: 0.971 g/cm3)
  • Color: light yellow to brown
  • Structure classification: ketones, aldehydes, acids

Catalog No. 50-004-54093 Supplier Medchemexpress LLC Supplier No. HYW0130145G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methyl-2-cyclopenten-1-one is an endogenous metabolite.

  • Endogenous metabolite
  • Appearance: liquid (density: 0.971 g/cm3)
  • Color: light yellow to brown
  • Structure classification: ketones, aldehydes, acids

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.