Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-heptene (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. M098901ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M098901ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M098901ML Supplier TCI America Supplier No. M09890.1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3404-75-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00143174
InChI Key OFKLSPUVNMOIJB-VMPITWQZSA-N
PubChem CID 5366149
IUPAC Name (2E)-3-methylhept-2-ene
SMILES CCCC\C(C)=C\C

Specifications

Color Colorless
Quantity 0.1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-heptene (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.