Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-nitroanisole, 98%, Thermo Scientific™

Catalog No. AAA1889003
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1889003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1889003 Supplier Thermo Scientific Chemicals Supplier No. A1889003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5345-42-6
Flash Point >110°C (230°F)
Molecular Formula C8H9NO3
Quantity 1g
MDL Number MFCD00007179
Beilstein 2329690
Synonym 3-methyl-2-nitroanisole, 3-methoxy-2-nitrotoluene, 2-nitro-3-methylanisole, benzene, 1-methoxy-3-methyl-2-nitro, pubchem12903, acmc-209l7h, anisole, 3-methyl-2-nitro, 3-methoxy-1-methyl-2-nitrobenzene, #
InChI Key MGBRGNWARSQECY-UHFFFAOYSA-N
SMILES COC1=CC=CC(C)=C1[N+]([O-])=O
IUPAC Name 1-methoxy-3-methyl-2-nitrobenzene
Molecular Weight (g/mol) 167.16
PubChem CID 79291
Formula Weight 167.16
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-nitroanisole
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 226-295-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.