Learn More
Medchemexpress LLC 3-Methyl-2-oxobutanoic acid-13C4,d4 sodium | 1185115-88-1 | 146.09 | 10 MG
SDP
Catalog No. 5000447460
Encompass_Preferred
Sodium 3-methyl-2-oxobutanoate-13C4,d4 is a deuterium and 13C labeled form of Sodium 3-methyl-2-oxobutanoate. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium and 13C labeled compound
  • Used as tracers for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-47460 Supplier Medchemexpress LLC Supplier No. HYW006057AS210MG
Sodium 3-methyl-2-oxobutanoate-13C4,d4 is a deuterium and 13C labeled form of Sodium 3-methyl-2-oxobutanoate. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium and 13C labeled compound
  • Used as tracers for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs