Learn More
Medchemexpress LLC 3-Methyl-2-oxobutanoic acid | 759-05-7 | 116.12 | 1 ML
SDP
Catalog No. 5000446278
Encompass_Preferred
3-Methyl-2-oxobutanoic acid is an endogenous metabolite and laboratory chemical used in research and the manufacture of substances. It acts as a precursor of pantothenic acid in Escherichia coli. This product typically presents as a colorless to off-white solid below 31.5°C and as a liquid above this temperature.
  • Functions as an endogenous metabolite
  • Serves as a precursor of pantothenic acid
  • Intended for research use only
  • Purity of 99.1% (LCMS)
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Harmful if swallowed
Catalog No. 50-004-46278 Supplier Medchemexpress LLC Supplier No. HYW00605710MM/1ML
3-Methyl-2-oxobutanoic acid is an endogenous metabolite and laboratory chemical used in research and the manufacture of substances. It acts as a precursor of pantothenic acid in Escherichia coli. This product typically presents as a colorless to off-white solid below 31.5°C and as a liquid above this temperature.
  • Functions as an endogenous metabolite
  • Serves as a precursor of pantothenic acid
  • Intended for research use only
  • Purity of 99.1% (LCMS)
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Harmful if swallowed