Learn More
Medchemexpress LLC 3-Methyl-2-oxobutanoic acid | 759-05-7 | 116.12 | 250 MG
SDP
Catalog No. 5000450079
Encompass_Preferred
3-Methyl-2-oxobutanoic acid is a precursor of pantothenic acid in *Escherichia coli*. This endogenous metabolite is classified under ketones, aldehydes, and acids, and serves as a disease marker for endocrine diseases and nervous system disorders. It is stable under recommended storage conditions.
  • Precursor of pantothenic acid in *Escherichia coli*
  • Endogenous metabolite
  • Classified under ketones, aldehydes, and acids
  • Disease marker for endocrine diseases and nervous system disorders
  • Stable under recommended storage conditions
  • Colorless to off-white appearance
  • Melting/freezing point: 31.5°C
  • Boiling point/range: 170.2°C at 760 mmHg
Catalog No. 50-004-50079 Supplier Medchemexpress LLC Supplier No. 250MGHYW006057R
3-Methyl-2-oxobutanoic acid is a precursor of pantothenic acid in *Escherichia coli*. This endogenous metabolite is classified under ketones, aldehydes, and acids, and serves as a disease marker for endocrine diseases and nervous system disorders. It is stable under recommended storage conditions.
  • Precursor of pantothenic acid in *Escherichia coli*
  • Endogenous metabolite
  • Classified under ketones, aldehydes, and acids
  • Disease marker for endocrine diseases and nervous system disorders
  • Stable under recommended storage conditions
  • Colorless to off-white appearance
  • Melting/freezing point: 31.5°C
  • Boiling point/range: 170.2°C at 760 mmHg