Learn More
Medchemexpress LLC 3-Methyl-2-oxovaleric acid | 1460-34-0 | 99.7% | 130.14 | 1 ML
SDP
Catalog No. 5000437305
Encompass_Preferred
3-Methyl-2-oxovaleric acid is a degradation product from Isoleucine. It is identified as a biomarker for mustard airway diseases (MADs) and uric acid stone.
  • Degradation product from Isoleucine
  • Biomarker for mustard airway diseases (MADs) and uric acid stone
  • Functions as a human endogenous metabolite
  • Levels are elevated in uric acid stone urinary environments
Catalog No. 50-004-37305 Supplier Medchemexpress LLC Supplier No. HY11306310MM/1ML
3-Methyl-2-oxovaleric acid is a degradation product from Isoleucine. It is identified as a biomarker for mustard airway diseases (MADs) and uric acid stone.
  • Degradation product from Isoleucine
  • Biomarker for mustard airway diseases (MADs) and uric acid stone
  • Functions as a human endogenous metabolite
  • Levels are elevated in uric acid stone urinary environments