Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methyl-2-oxovaleric acid sodium | 3715-31-9 | MFCD00002584 | 99.6% | 152.12 g/mol | C6H9NaO3 | 250 MG
SDP

Catalog No. 5000303745
Encompass_Preferred

3-methyl-2-oxovaleric acid sodium is the sodium salt of 3-methyl-2-oxovaleric acid, an endogenous metabolite and degradation product of isoleucine. It is used as a research reagent and reference standard and is reported as a biomarker for mustard airway diseases and uric acid stones.

  • High purity, 99.6%.
  • Molecular formula C6H9NaO3; molecular weight 152.12 g/mol.
  • Solid, white to off-white appearance.
  • Soluble in DMSO at 50 mg/mL; ultrasonic aid recommended.
  • Store sealed, away from moisture; in solution: -80°C (6 months), -20°C (1 month).
  • Used as a reference standard and in studies of α-keto acid metabolism.

Catalog No. 50-003-03745 Supplier Medchemexpress LLC Supplier No. 250MGHYW017386
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-methyl-2-oxovaleric acid sodium is the sodium salt of 3-methyl-2-oxovaleric acid, an endogenous metabolite and degradation product of isoleucine. It is used as a research reagent and reference standard and is reported as a biomarker for mustard airway diseases and uric acid stones.

  • High purity, 99.6%.
  • Molecular formula C6H9NaO3; molecular weight 152.12 g/mol.
  • Solid, white to off-white appearance.
  • Soluble in DMSO at 50 mg/mL; ultrasonic aid recommended.
  • Store sealed, away from moisture; in solution: -80°C (6 months), -20°C (1 month).
  • Used as a reference standard and in studies of α-keto acid metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.