Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-pentanone, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
565-61-7
C6H12O
100.16
UIHCLUNTQKBZGK-UHFFFAOYSA-N
3-methyl-2-pentanone,2-pentanone, 3-methyl,methyl sec-butyl ketone,sec-butyl methyl ketone,methyl 1-methylpropyl ketone,+/--3-methyl-pentan-2-one,methylethylacetone,sec-c4h9coch3,3-methyl-pentan-2-one,acmc-1alw4
11262
3-methylpentan-2-one
CCC(C)C(=O)C
565-61-7
C6H12O
100.16
UIHCLUNTQKBZGK-UHFFFAOYSA-N
3-methyl-2-pentanone,2-pentanone, 3-methyl,methyl sec-butyl ketone,sec-butyl methyl ketone,methyl 1-methylpropyl ketone,+/--3-methyl-pentan-2-one,methylethylacetone,sec-c4h9coch3,3-methyl-pentan-2-one,acmc-1alw4
11262
3-methylpentan-2-one
CCC(C)C(=O)C
Specific Gravity 0.81
Flash Point 12°C
Boiling Point 118.0°C (758.0 mmHg)
Refractive Index 1.4002
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Density 0.8100g/mL
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C2H5CH(CH3)COCH3
Beilstein 01,693
Formula Weight 100.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Methyl-2-pentanone
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapour.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger