Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-quinoxalinol, 98%, Thermo Scientific™

Catalog No. AC162970250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162970250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162970250 Supplier Thermo Scientific Chemicals Supplier No. 162970250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14003-34-0
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00006724
InChI Key BMIMNRPAEPIYDN-UHFFFAOYSA-N
Synonym 3-methylquinoxalin-2-ol, 3-methyl-2-quinoxalinol, 2-hydroxy-3-methylquinoxaline, 3-methylquinoxalin-2 1h-one, 2-quinoxalinol, 3-methyl, usaf el-7, 2 1h-quinoxalinone, 3-methyl, 3-methyl-2 1h-quinoxalinone, 3-hydroxy-2-methylquinoxaline, 2-hydroxy-3-methyl quinoxaline
PubChem CID 26384
IUPAC Name 3-methyl-1H-quinoxalin-2-one
SMILES CC1=NC2=CC=CC=C2NC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 246°C to 248°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 24, 165
Formula Weight 160.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Methyl-2-quinoxalinol, 98%
Show More Show Less
EINECSNumber 237-807-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.