Learn More
Medchemexpress LLC 3-[methyl[3-[4-(phenylmethyl)phenoxy]propyl]amino]propanoic acid HCl | 423169-68-0 | MFCD00951253 | 99.9% | 363.88 | C20H26ClNO3 | 50 MG
SDP
Catalog No. 5000246531
Encompass_Preferred
SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase used for research. It shows low-nanomolar inhibitory activity against human, mouse, and rat LTA4 hydrolase and is supplied as a purified solid for laboratory studies.
  • Potent LTA4 hydrolase inhibition with low-nanomolar IC50s.
  • Intended for research use only, not for human or veterinary use.
  • High purity (≈99.9%).
  • White to off-white solid appearance.
  • Molecular weight 363.88; formula C20H26ClNO3.
  • Supplied as a 50 mg vial suitable for bench-scale experiments.
Catalog No. 50-002-46531 Supplier Medchemexpress LLC Supplier No. HY10322650MG
SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase used for research. It shows low-nanomolar inhibitory activity against human, mouse, and rat LTA4 hydrolase and is supplied as a purified solid for laboratory studies.
  • Potent LTA4 hydrolase inhibition with low-nanomolar IC50s.
  • Intended for research use only, not for human or veterinary use.
  • High purity (≈99.9%).
  • White to off-white solid appearance.
  • Molecular weight 363.88; formula C20H26ClNO3.
  • Supplied as a 50 mg vial suitable for bench-scale experiments.