Learn More
Medchemexpress LLC 3-[methyl[3-[4-(phenylmethyl)phenoxy]propyl]amino]propanoic acid HCl | 423169-68-0 | MFCD00951253 | 99.9% | 363.88 g·mol⁻¹ | C20H26ClNO3 | 10 MG
SDP
Catalog No. 5000246809
Encompass_Preferred
SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase used for research into inflammatory pathways. It exhibits low-nanomolar inhibitory activity against human, mouse, and rat LTA4 hydrolase and is supplied as a solid powder with high purity and defined storage recommendations.
  • Potent LTA4 hydrolase inhibition with low-nanomolar IC50 values.
  • High purity suitable for research applications.
  • Supplied as a white to off-white solid powder for ease of handling.
  • Stable under recommended storage conditions to preserve activity.
  • Available in small-mass packages suitable for exploratory studies.
Catalog No. 50-002-46809 Supplier Medchemexpress LLC Supplier No. HY10322610MG
Add to Cart
Add to Cart
SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase used for research into inflammatory pathways. It exhibits low-nanomolar inhibitory activity against human, mouse, and rat LTA4 hydrolase and is supplied as a solid powder with high purity and defined storage recommendations.
  • Potent LTA4 hydrolase inhibition with low-nanomolar IC50 values.
  • High purity suitable for research applications.
  • Supplied as a white to off-white solid powder for ease of handling.
  • Stable under recommended storage conditions to preserve activity.
  • Available in small-mass packages suitable for exploratory studies.