Learn More
Medchemexpress LLC 3-[methyl[3-[4-(phenylmethyl)phenoxy]propyl]amino]propanoic acid hydrochloride | 423169-68-0 | MFCD00951253 | 99.9% | 363.88 | C20H26ClNO3 | 100 MG
SDP
Catalog No. 5000246712
Encompass_Preferred
SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase intended for research use. It shows low-nanomolar activity against recombinant human, mouse, and rat LTA4 hydrolase and is supplied as a solid or as DMSO solution for laboratory experiments.
  • Potent LTA4 hydrolase inhibitor with low-nanomolar IC50s.
  • High purity suitable for research assays.
  • Molecular weight 363.88 and formula C20H26ClNO3.
  • Available as a 100 mg solid and as DMSO solutions.
  • Intended for research use only.
Catalog No. 50-002-46712 Supplier Medchemexpress LLC Supplier No. HY103226100MG
SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase intended for research use. It shows low-nanomolar activity against recombinant human, mouse, and rat LTA4 hydrolase and is supplied as a solid or as DMSO solution for laboratory experiments.
  • Potent LTA4 hydrolase inhibitor with low-nanomolar IC50s.
  • High purity suitable for research assays.
  • Molecular weight 363.88 and formula C20H26ClNO3.
  • Available as a 100 mg solid and as DMSO solutions.
  • Intended for research use only.