Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methyl-3-buten-1-ol

Catalog No. 129402100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-940-2100ML 100 mL
12-940-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-940-2100ML Supplier Sigma Aldrich Supplier No. 129402100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Methyl-3-buten-1-ol was used to study the rate coefficient for the gas-phase reaction of OH radical with 3-methyl-3-buten-1-ol.

Specifications

CAS 763-32-6
Density 0.853 g/mL (at 25°C (literature))
Boiling Point 130°C to 132°C (lit.)
Molecular Formula C5H10O
Refractive Index n20/D 1.433 (literature)
Linear Formula CH2=C(CH3)CH2CH2OH
MDL Number MFCD00002933
Quantity 100 mL
Molecular Weight (g/mol) 86.13
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.