Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methyl-3H-imidazo[4,5-b]pyridin-6-ol

Catalog No. ADE0012231G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0012231G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0012231G Supplier Sigma Aldrich Supplier No. ADE0012231G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-fluorobenzenesulfonyl chloride, also known as m-fluorobenzenesulfonyl chloride, is a fluorinated arylsulfonyl chloride
  • It can be prepared from 1,3-benzenedisulfonyl fluoride.3-fluorobenzenesulfonyl chloride may be used in the preparation of 1-(2-bromobenzyl)-2-(3-fluorophenyl)pyrrole and 1-(2-bromobenzyl)-2,5-bis(3-fluorophenyl)pyrrole

Specifications

CAS 1171920-70-9
Molecular Formula C7H7N3O
Linear Formula C7H7N3O
Quantity 5 g
MDL Number MFCD12401667
Molecular Weight (g/mol) 149.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.