Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-4-nitrobenzoic acid, 99%

Catalog No. AAA1552336
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1552336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1552336 Supplier Thermo Scientific Chemicals Supplier No. A1552336

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 3113-71-1 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3113-71-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007168
InChI Key XDTTUTIFWDAMIX-UHFFFAOYSA-N
PubChem CID 18370
SMILES CC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 215°C to 220°C
Quantity 500 g
Beilstein 1959154
Synonym 3-methyl-4-nitrobenzoic acid, 4-nitro-m-toluic acid, benzoic acid, 3-methyl-4-nitro, m-toluic acid, 4-nitro, unii-elp258jxh0, 3-methyl-4-nitrobenzoicacid, 3-methyl-4-nitro-benzoic acid, elp258jxh0, benzoic acid,3-methyl-4-nitro, 3-methyl-4-nitro benzoic acid
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 3-Methyl-4-nitrobenzoic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 221-479-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.