Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-4-nitrothiobenzamide, 97%, Thermo Scientific™

Catalog No. AAH5455003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5455003 1 g
AAH54550MD 250 mg
AAH5455006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5455003 Supplier Thermo Scientific Chemicals Supplier No. H5455003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886364-22-3
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
MDL Number MFCD06738330
InChI Key QNPFIXDGSZDAGC-UHFFFAOYSA-N
Synonym 3-methyl-4-nitrothiobenzamide, 3-methyl-4-nitrobenzothioamide, 3-methyl-4-nitrobenzene-1-carbothioamide
PubChem CID 55255791
IUPAC Name 3-methyl-4-nitrobenzenecarbothioamide
SMILES CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]

Specifications

Quantity 1 g
Beilstein 19566115
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrothiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.