Learn More
Medchemexpress LLC 3-methyl-5-(1-methyl-2-pyrrolidinyl)isoxazole hydrochloride | 147388-83-8 | MFCD11045960 | 99.4% | 202.68 g·mol⁻¹ | C9H15ClN2O | 100 MG
SDP
Catalog No. 5000243400
Encompass_Preferred
ABT-418 hydrochloride is the hydrochloride salt of a selective neuronal nicotinic acetylcholine receptor (nAChR) agonist used in preclinical research to study cognition and anxiety-related pharmacology. It is provided as a high-purity solid suitable for analytical, in vitro, and in vivo experiments.
  • Hydrochloride salt of a selective neuronal nicotinic acetylcholine receptor agonist.
  • Intended for in vitro and in vivo pharmacology and neuroscience research.
  • High purity (99.4%) suitable for analytical and pharmacological studies.
  • Molecular weight 202.68 g·mol⁻¹ and formula C9H15ClN2O.
  • Available as a solid in multiple package sizes, including 100 mg.
Catalog No. 50-002-43400 Supplier Medchemexpress LLC Supplier No. HY105170B100MG
ABT-418 hydrochloride is the hydrochloride salt of a selective neuronal nicotinic acetylcholine receptor (nAChR) agonist used in preclinical research to study cognition and anxiety-related pharmacology. It is provided as a high-purity solid suitable for analytical, in vitro, and in vivo experiments.
  • Hydrochloride salt of a selective neuronal nicotinic acetylcholine receptor agonist.
  • Intended for in vitro and in vivo pharmacology and neuroscience research.
  • High purity (99.4%) suitable for analytical and pharmacological studies.
  • Molecular weight 202.68 g·mol⁻¹ and formula C9H15ClN2O.
  • Available as a solid in multiple package sizes, including 100 mg.